Order Entry
Canada
ContactUsLinkComponent
Human Recombinant alpha-Synuclein (from E. coli), Biotin
Human Recombinant alpha-Synuclein (from E. coli), Biotin
Catalog # 103001-712
Supplier:  Anaspec Inc
CAS Number:  
Human Recombinant alpha-Synuclein (from E. coli), Biotin
Catalog # 103001-712
Supplier:  Anaspec Inc
Supplier Number:  AS-55581
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Biotin
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    Store at 2-4 °C for immediate use within 1 week or at –80 °C for up to 12 months. Keep in dark and avoid repeated freeze-thaw cycles.
  • Gene ID:
    NP_000336
  • Protein synonyms:
    Synuclein-HiLyte™ Fluor 488|Synuclein 1-140 HF 489|α-Synuclein (1-140) labeled|Synuclein-HF 488
  • Protein/peptide name:
    alpha-Synuclein
  • Purity:
    > 95% as determined by SDS-PAGE
  • Cat. no.:
    103001-712
  • Supplier No.:
    AS-55581

Specifications

About this item

The recombinant human α-synuclein (1-140) (GenBank Accession # NP_000336) was expressed and purified from E. coli and conjugated with biotin. Recombinant protein is produced without an affinity tag.
α-Synuclein is a major component of Lewy bodies in the affected neurons in Parkinson's disease. This protein has a mass of 14.5 kDa (140 amino acids long) and consists of a conserved degenerative amino-terminal domain and an acidic carboxyl-terminal with higher sequence divergence. α-Synuclein is predominantly expressed in brain, specifically in cerebellum, thalamus, neocortex, hippocampus, and striatum regions. Other tissues express α-Synuclein at very low levels. The physiological role of α-synuclein is not yet well understood. However, the presence of imperfect KTKEGV lipid interacting repeats suggests that it may be involved in synaptic vesicle homeostasis.