Order Entry
Canada
ContactUsLinkComponent
Human Recombinant alpha-Synuclein (from E. coli), HiLyte Fluor® 488
Human Recombinant alpha-Synuclein (from E. coli), HiLyte Fluor® 488
Catalog # 103004-984
Supplier:  Anaspec Inc
CAS Number:  
Human Recombinant alpha-Synuclein (from E. coli), HiLyte Fluor® 488
Catalog # 103004-984
Supplier:  Anaspec Inc
Supplier Number:  AS-55457
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    HiLyte Fluor® 488
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    200 µg
  • Storage conditions:
    Store at 2-4 °C for immediate use within 1 week or at –80 °C for up to 12 months. Keep in dark and avoid repeated freeze-thaw cycles.
  • Gene ID:
    NP_000336
  • Protein synonyms:
    Synuclein-HiLyte™ Fluor 488|α-Synuclein (1-140) labeled|Synuclein 1-140 HF 488|Synuclein-HF 488
  • Protein/peptide name:
    alpha-Synuclein
  • Purity:
    > 95% as determined by SDS-PAGE
  • Cat. no.:
    103004-984
  • Supplier No.:
    AS-55457

Specifications

About this item

The recombinant human a-synuclein (1-140) (GenBank Accession # NP_000336) was expressed and purified from E. coli and conjugated with the fluorescence dye HiLyte FluorTM 488. This protein is produced without affinity tag.
α-Synuclein is a major component of Lewy bodies in the affected neurons in Parkinson's disease. This protein has a mass of 14.5 kDa (140 amino acids long) and consists of a conserved degenerative amino-terminal domain and an acidic carboxyl-terminal with higher sequence divergence. α-Synuclein is predominantly expressed in brain, specifically in cerebellum, thalamus, neocortex, hippocampus, and striatum regions. Other tissues express α-Synuclein at very low levels. The physiological role of α-synuclein is not yet well understood. However, the presence of imperfect KTKEGV lipid interacting repeats suggests that it may be involved in synaptic vesicle homeostasis.