BBL™ Phosphate Buffer, BD
Phosphate buffer, 500 ml, 6.8
Supplier: BD
Supplier #: 297657
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This is used as buffered diluent for microbiological procedures.
Documents
Specifications
Specifications
- Description:Phosphate buffer
- Size:500 ml
- pH:6.8
- Cat. No.:CA90004-948
- Shelf life:365



