Order Entry
Canada
ContactUsLinkComponent
Trastuzumab ELISA Kit, IBL America
Trastuzumab ELISA Kit, IBL America
Catalog # 102978-466
Supplier:  IBL-America
CAS Number:  
Trastuzumab ELISA Kit, IBL America
Catalog # 102978-466
Supplier:  IBL-America
Supplier Number:  TM09013
CAS Number:  

Specifications

  • Conjugate ELISA:
    HRP
  • Primary antibody reactivity:
    Human
  • Target protein:
    Trastuzumab
  • Description:
    Trastuzumab ELISA kit
  • Size:
    96 Determinations, 12x8 Removable Strips
  • Sample type:
    Serum and Plasma
  • Detection range:
    0.6 - 200 ng/mL
  • Storage temperature:
    2...8 °C
  • Sensitivity:
    5 ng/mL
  • Cat. no.:
    102978-466
  • Supplier No.:
    IG-AA105

Specifications

About this item

ELISA test kit for the determination of free Trastuzumab (trade name Herceptin®, Herclon®) in serum and plasma

Trastuzumab is a recombinant DNA-derived humanized monoclonal antibody that selectively targets the extracellular domain of the human epidermal growth factor receptor 2 protein (HER2).

The catcher is a mixture of HER2/ErbB2/CD340 and the detection antibody is a HRP-labeled mAb binding at the Fc part of the drug.