Order Entry
Canada
ContactUsLinkComponent
Terrific Broth Complete with 100 µg/ml Carbenicillin Plate Media, Teknova
Terrific Broth Complete with 100 µg/ml Carbenicillin Plate Media, Teknova
Catalog # CA100217-078
Supplier:  Teknova
CAS Number:  
Terrific Broth Complete with 100 µg/ml Carbenicillin Plate Media, Teknova
Catalog # CA100217-078
Supplier:  Teknova
Supplier Number:  T4978
CAS Number:  

Specifications

  • Description:
    Terrific Broth Complete with 100 µg/mL Carbenicillin
  • Sterility:
    Sterile
  • Cat. no.:
    CA100217-078
  • Plate Diameter:
    150 mm
  • Supplier No.:
    T4978

Specifications

About this item

Carbenicillin is an analog of ampicillin

  • Sleeve of 20 plates
  • Sterile

Ingredients: 1.2% Bacto Tryptone, 2.5% Yeast Extract, 1.6% glycerol, 1.5% Agar, 100 µg/mL Carbenicillin, 72 mM K2HPO4, and 17 mM KH2PO4.

Ambient shipping conditions.

For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids.