Orders LinkContactUsLinkComponent
Spring Scales
Spring Scales
Supplier:   Ohaus
Spring Scales
Supplier:  Ohaus
Certificates
4 Options Available Below
Product Details & Documents
Ideal for general weighing, force experiments, action-reaction demonstrations.
  • Dial plates display
  • Metric/ounce/newtons
Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.
Product Family Options
Pully type
Product Information
  • Weighing capacity
    Availability
    Price
  • 89425-988
    8001-MN - 10 g (0.1 N) graduation intervals
    8001-MN - 10 g (0.1 N) graduation intervals
    250 g (2.5 N)
    Log in for availability
    Log in for availability
    Each
    Each - $60.71
    $60.71
    Each
       $60.71
    Supplier Number: 80000014
  • 89425-994
    8002-MN - 20 g (0.2 N) graduation intervals
    8002-MN - 20 g (0.2 N) graduation intervals
    500 g (5 N)
    Log in for availability
    Log in for availability
    Each
    Each - $60.71
    $60.71
    Each
       $60.71
    Supplier Number: 80000015
  • 89426-004
    8004-MN - 50 g (0.5 N) graduation intervals
    8004-MN - 50 g (0.5 N) graduation intervals
    2000 g (20 N)
    Log in for availability
    Log in for availability
    Each
    Each - $60.71
    $60.71
    Each
       $60.71
    Supplier Number: 80000017
  • 97027-720
    8001-MA - 10 g (0.25 oz) graduation intervals
    8001-MA - 10 g (0.25 oz) graduation intervals
    250 g (9 oz.)
    Log in for availability
    Log in for availability
    Each
    Each - $58.18
    $58.18
    Each
       $58.18
    Supplier Number: 80000039