Order Entry
Canada
Orders LinkContactUsLinkComponent
Multi-Element Quality Control Standard, 21 Elements, PerkinElmer

Multi-Element Quality Control Standard, 21 Elements, PerkinElmer

Quality Control Standard, 21 Elements, 125 ml, 5% HNO₃
Catalog # CA97011-998
Supplier:   PerkinElmer
Supplier #:  N9300281
Multi-Element Quality Control Standard, 21 Elements, PerkinElmer
Catalog # CA97011-998
Supplier:  PerkinElmer
CAS Number:  
Multi-Element Quality Control Standard, 21 Elements, PerkinElmer
Multi-Element Quality Control Standard, 21 Elements, PerkinElmer

Quality Control Standard, 21 Elements, 125 ml, 5% HNO₃

CAS Number:  
Price
$864.06
Log in for availability
Each (100ml)
Product Details & Documents

Use for standard metal analysis.

  • Matrix: 5% HNO3
  • Liquid format

21 elements (100 mg/L): As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V and Zn.

Documents

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Description:
    Quality Control Standard, 21 Elements
  • Size:
    125 ml

Specifications