Trypsin Neutralizing Solution, PromoCell
About this item
Trypsin is the most widely employed enzyme used for cell detachment
This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0.05% trypsin inhibitor in 0.1% BSA.
Caution: For in vitro research use only and not for diagnostic or therapeutic procedures.
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Trypsin is the most widely employed enzyme used for cell detachment
This solution exhibit standardized enzyme activity and contain trypsin from extra-pure lots. This solution contains 0.05% trypsin inhibitor in 0.1% BSA.
Caution: For in vitro research use only and not for diagnostic or therapeutic procedures.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10172-556
- 10175-288
- 10175-290
- Storage temperature:
- –20 °C
- –20 °C
- –20 °C
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 10172-556Trypsin Neutralizing SolutionSupplier Number: C-41100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10172-556Storage temperature–20 °C - 10175-288Trypsin Neutralizing SolutionSupplier Number: C-41110Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10175-288Storage temperature–20 °C - 10175-290Trypsin Neutralizing SolutionSupplier Number: C-41120Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10175-290Storage temperature–20 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



