Order Entry
Canada
Orders LinkContactUsLinkComponent
EMB Agar Plates

EMB Agar Plates

Supplier:   Teknova
EMB Agar Plates
Supplier:  Teknova
Certificates

2 Options Available Below

Product Details & Documents

EMB (Eosin Blue) agar plates are used as a slightly selective and differential plating medium for the isolation of gram-negative enteric bacteria

  • Sterile
  • Final pH = 7.1 ±10.2 (at 25 °C)

These plates are commonly used as bacterial indicator plates to monitor fermentation. It contains 1.0% Peptone (10.0 g/L), 2.0% Lactose (20.0 g/L), 0.2% Potassium Phosphate, K2HP04 (5.0 g/L), 0.04% Eosin Y (0.4 g/L), 0.0065% Methylene Blue (0.065 g/L), 1.5% Agar (15.0 g/L).

Product Family Options

Product Information

  • Plate diameter
    Size
    Availability
    Price
  • CA100219-774
    EMB (Eosin Blue) Agar
    EMB (Eosin Blue) Agar
    100 mm
    20 Plates
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $834.82
    $834.82
    Pack of 20
       $834.82
    Minimum order quantity is 8

    Supplier Number: E0070
  • CA100219-680
    EMB (Eosin Blue) agar
    EMB (Eosin Blue) agar
    150 mm
    20 Plates
    Discontinued
    Supplier Number: -
  • CA77443-426
    EMB (Eosin Blue) agar
    EMB (Eosin Blue) agar
    100 mm
    20 Plates
    Log in for availability
    Log in for availability
    Each
    Each - $212.23
    $212.23
    Each
       $212.23
    Minimum order quantity is 1

    Supplier Number: E0070
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.