Order Entry
Canada
ContactUsLinkComponent
SensoLyte® 520 Cathepsin D Assay Kit Fluorimetric, AnaSpec, Inc.
SensoLyte® 520 Cathepsin D Assay Kit Fluorimetric, AnaSpec, Inc.
Catalog # 103010-544
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 Cathepsin D Assay Kit Fluorimetric, AnaSpec, Inc.
Catalog # 103010-544
Supplier:  Anaspec
Supplier Number:  AS-72170
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 Kit
  • Storage temperature:
    -20°C
  • Cat. no.:
    103010-544
  • No. of Wells:
    96
  • No. of Assays/Kit:
    100 assays
  • Supplier No.:
    AS-72170

Specifications

About this item

Cathepsin D is the lysosomal aspartic proteinase, active in intracellular protein breakdown

Cathepsins are a class of globular lysosomal proteases playing a vital role in mammalian cellular turnover. They degrade polypeptides and are distinguished by their substrate specificities.
The cathepsin assay kits offers a series of fluorimetric long wavelength, sensitive SensoLyte® 520 assay kits for enzyme activity determination of Cathepsins G, D, E, B, K, L and S. These kits employ a FRET (Fluorescence Resonance Energy Transfer) peptide substrate that emits in the green range of 520 nm, with excitation at 490 nm. The advantage of using longer wavelength emitting substrates means that autofluorescence from cellular components and test compounds is minimal; thus making the kits more sensitive. Furthermore, the use of HiLyte Fluor™ 488 (except for Cathepsin S, which uses 5-FAM) as the FRET donor (fluorophore) results in a more stable fluorescent signal, since HiLyte Fluor™ 488 is resistant to acidic pH. A listing of the SensoLyte® Cathepsin Kits providing conventional FRET (Mca/Dnp) or fluorogenic substrates for cathepsins are also available.

Kits detect specific Cathepsin activities; provides ample reagents to perform 100 assays in 96-well format; minimal hands-on time required; detailed protocol and references provided.

Cathepsin D is involved in the pathogenesis of several diseases such as breast cancer and Alzheimer disease