If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:SensoLyte® pNPP Protein Phosphatase Assay Kit *Colorimetric*
- Size:1 Kit
- Storage temperature:- 20 °C
- Cat. no.:103010-118
- No. of Wells:96
- No. of Assays/Kit:500 assays (96 well plate) or 1250 assays (384 well plate)
- Supplier No.:AS-71105
Specifications
About this item
p-Nitrophenyl phosphate (pNPP) is proven to be an effective chromogenic substrate for protein tyrosine phosphatases and serine/threonine phosphatases
Protein phosphatases have received great attention as drug-screening targets. SensoLyte® Protein Phosphatase Assay Kits uses specific substrates to quantify specific protein phosphatase activities such as MFP, pNPP FDP or acid phosphatases. The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of protein phosphatase inhibitors/inducers.
Optimized to detect protein phosphatase activity; compatible with HTS; provides enough reagents to perform 500 assays in a 96-well format; minimal hands-on time; detailed protocol and references provided.
SensoLyte® pNPP Protein Phosphatase Assay Kit uses pNNP to quantify protein phosphatase activities. The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of protein phosphatase inhibitors. It has high sensitivity and wide linear range (The detection limit is generally 3 ng or below).