Stroboscope
Stroboscope
470230-254
: Wards
Supplier #: PH0752A
Sturdy stroboscope ideal for studying periodic motion.
- 9.5 inch diameter rotating disc on handle
- Total height: 18 inches
- Sturdy plastic disc with slots
- Disc includes finger hold
- Handle with rubber grip
The sturdy stroboscope with marked disc, finger hold in disc, and rubber gripped handle is easy to use and ideal for studying periodic motion and speed of rotation.
- Description:Stroboscope