Spectrometer Advanced
Spectrometer, Advanced
Catalog # 470230-220
Supplier: Wards
Supplier #: PH0620
Jump to:
Product Details & Documents
Spectrometer for studying concepts of optics.
- Includes: Spectrometer, 1 prism holder, and 1 grating holder
- Solid base with turntable, prism table, collimator, and telescope
- Used to determine wavelength of light
- Used to determine the index of refraction of solids and liquids
- Stored in polished wooden case
The advanced spectrometer, including 1 prism holder and 1 grating holder has a solid base with a turntable, prism table, collimator, and telescope, is ideal for determining both the wavelength of light and the index of refraction of solids and liquids.
Specifications
- Description:Spectrometer, Advanced
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.