Order Entry
Canada
ContactUsLinkComponent
Bimetallic Strips on Blocks, Set 10
Bimetallic Strips on Blocks, Set 10
Catalog # 470230-164
Supplier:  Wards
undefined
Bimetallic Strips on Blocks, Set 10
Catalog # 470230-164
Supplier:  Wards
Supplier Number:  PH0382

Specifications

  • Description:
    BiMetallic Strips on Blocks, Set 10
  • Cat. no.:
    470230-164

Specifications

About this item

Bimetallic strips attached to wood blocks to study thermal energy.

  • 10 strips per pack
  • Bimetallic strip attached to wood block (block 1.85 x 1.45 x 0.4")
  • Total length of item: 4.31"
  • Strips attached by 2 nails

Bimetallic strips are sturdy strips of two metals attached to solid wood blocks and can be used to study thermal energy converting to mechanical motion (this is caused by the different expansions of two different metals when heated).