ContactUsLinkComponent
Double Cone and Plane
Double Cone and Plane
Catalog # 470230-148
Supplier:  Wards
undefined
Double Cone and Plane
Catalog # 470230-148
Supplier:  Wards
Supplier Number:  PH0349DCP
Specifications
  • Description:
    Double Cone and Plane
  • Cat. no.:
    470230-148
Specifications
About this item
Demonstrates center of mass with wooden plane and double cone that comes assembled.
  • Center of mass demonstration
  • Apparatus comprises of a wooden A from with diverging rails and a double ended cone
  • Easy use, place cone of the bottom of the ramp and let go
  • Constructed of durable wood and clear coated for decades of use
  • Comes assembled
This is a great toy, executive gift, or physics demonstration as the cone appears to be traveling "Up" the ramp. The physics behind the demonstration is that as the double-ended cone moves up the ramp the center of mass is actually moving downward which drives the cone up the ramp itself. Amaze your students and colleagues.