Order Entry
Canada
ContactUsLinkComponent
Hydrochloric acid 2%, Claritas PPT® Grade for calibration blank in ICP
Hydrochloric acid 2%, Claritas PPT® Grade for calibration blank in ICP
Catalog # SXCLBLK-HCL
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Hydrochloric acid 2%, Claritas PPT® Grade for calibration blank in ICP
Catalog # SXCLBLK-HCL
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CLBLK-HCL
CAS Number:  

Specifications

  • Pack type:
    LDPE bottle
  • Size:
    125 ml
  • Cat. no.:
    SXCLBLK-HCL

Specifications

About this item

 :
ADR:8,III
 :36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324