Hooke's Law Apparatus with Weights
Catalog # 470230-118
Supplier: Wards
Specifications
- Description:Hooke's Law Apparatus with Weights
- Cat. no.:470230-118
Specifications
About this item
Hooke's Law apparatus, includes weights for demonstration.
- 15 cm adjustable scale
- Scale marked in mm
- Includes experimental notes
- Wooden base
This complete apparatus has a 15cm adjustable scale. The adjustable scale helps to prevent parallax error. Scale is marked in millimeters and mounted on sturdy 30 cm support rod and a wooden base. A hook supporting a coiled spring with a mass hanger and indicator is attached to rod. 10 g weight plates included along with experimental notes.