Friction Block with Hook
Catalog # 470230-068
Supplier: Wards
Specifications
- Description:Friction Block with Hook
- Cat. no.:470230-068
Specifications
About this item
Friction blocks are useful for teaching the basics of force and motion.
- Wooden friction block with c-hook attached at the terminus
- Use with a spring scale to measure force and mass - all the numbers you need to solve a variety of motion and force problems
- Hands-on, economic tool for teaching fundamental concepts in physics
- Students of any age can learn using the friction block
- Manufactured in the US
This wooden friction block is perfect for introductory studies in force and motion. The block is fitted with a c-hook so that it can be attached to a spring scale. Slide the block along a flat surface, or pull it up an incline. Observe the block by itself, or add weights to the top. When used in a tandem with a spring scale, the friction block is a powerful tool for learning about friction, kinetic and potential energy, acceleration, and force. This block is sourced and manufactured in the United States.