Order Entry
Canada
ContactUsLinkComponent
Density Cubes with Case
Density Cubes with Case
Catalog # 470230-038
Supplier:  Wards
undefined
Density Cubes with Case
Catalog # 470230-038
Supplier:  Wards
Supplier Number:  PH0109A

Specifications

  • Description:
    Density Cubes with Case
  • Cat. no.:
    470230-038

Specifications

About this item

Set of 9 cubes, 25 mm side. Supplied in wooden box.

  • Density Block set consists of 9 blocks
  • 1" square blocks
  • Materials included brass, iron, copper, aluminum, hardwood, softwood, nylon, PVC and acrylic.
  • Supplied in 5.75" x 5.75" x 1.25" wooden box with hinges and clasp.
  • Perfect for student studying mass, volume and density

This set consists of nine cubes for density investigation, 25 mm side. Set of 9 comprising brass, iron, copper, aluminum, hardwood, softwood, nylon, PVC and acrylic. Supplied in wooden box.