Orders LinkContactUsLinkComponent
Neulog Pulley Kit
Neulog Pulley Kit
Neulog pulley kit
Catalog # 470229-978
Supplier:   Wards
Supplier #:  PLPSES
Neulog Pulley Kit
Catalog # 470229-978
Supplier:  Wards
Neulog Pulley Kit
Neulog Pulley Kit
Neulog pulley kit
Price
$101.50
Log in for availability
Kit of 1
Product Details & Documents
Hardware to conduct pulley experiments with NeuLog sensors.
  • Provides students with a visual and hands on learning experience!
  • Illustrates the differences between fixed a fixed pulley, a compound pulley and a moveable pulley
  • NOT INCLUDED: Neulog force sensor logger (item number NUL211)
  • Kit comes with comprehensive instructions and detailed experiment guide
  • 23 inch operational height, stable 16×6" base
This kit allows the user to study general properties of fixed pulleys, movable pulleys and compound pulleys. It requires the purchase of the Neulog force logger sensor (NUL211) and provides beautifully illustrative data that allows students to learn about the complexities and differences in different pulley times in a manner that combines visual learning with hands on experimenting.
Ordering information: Kit contains 35 cm track, 2 track legs, 2×20" rods, 2 track riders, 2 boss heads, 2×100g slotted masses rod with perpendicular pulley with hook thread instructions and detailed experiment guide.
Caution: For educational use only.
Specifications
  • Description:
    Neulog pulley kit