Celestron Flipview 5MP LCD Portable Microscope
Catalog #: 470229-742
Supplier: Celestron International
Irregular Voltage
Specifications
- Magnification:10x to 120x
- Description:5MP LCD Portable Microscope, 100 − 240 V
- Model:44314
- Cat. no.:470229-742
Specifications
About this item
Handheld digital microscope with integrated 3-inch full color TFT LCD screen that flips out and rotates 270 degrees.
- Versatile ergonomic design can be used three ways: handheld, on a flat surface with flip out leg, or with the included metal stand and 1/4-20 connection
- 5 MP sensor and 5-element IR cut lens configuration captures high definition images and video
- 10x to 120x magnification (up to 300x on PC)
- Rechargeable lithium ion battery
This compact yet powerful digital microscope is the perfect companion for up-close observation on the go. The onboard 3-inch, full color TFT LCD screen flips out when you need it, then folds back into the microscope body for portability. A rechargeable lithium ion battery provides hours of viewing time in the classroom or on a nature walk. FlipView, with its 10x to 120x magnification is a low-power microscope perfectly designed for viewing an array of 3D specimens, from beach sand to bugs, coins and currency, documents, circuit boards, and more.
Customers Who Bought This Also Bought