Order Entry
Canada
ContactUsLinkComponent
BD BBL™ Trypticase™ Soy Agar with Lecithin, Polysorbate 80, and Penicillinase Settling Plates, BD Diagnostics
BD BBL™ Trypticase™ Soy Agar with Lecithin, Polysorbate 80, and Penicillinase Settling Plates, BD Diagnostics
Catalog # CA-90004-196
Supplier:  BD
CAS Number:  
undefined
BD BBL™ Trypticase™ Soy Agar with Lecithin, Polysorbate 80, and Penicillinase Settling Plates, BD Diagnostics
Catalog # CA-90004-196
Supplier:  BD
Supplier Number:  292301
CAS Number:  

Specifications

  • Description:
    Trypticase™ Soy Agar with Lecithin, Polysorbate 80, and Penicillinase
  • Cat. no.:
    CA-90004-196
  • Plate Diameter:
    100 mm
  • Supplier No.:
    292301

Specifications

About this item

BD BBL™ prepared agar plates in sterile packs are plated media in gamma-irradiated packs that are validated to performance and sterility according to AAMI (Association for the Advancement of Medical Instrumentation) guidelines

  • For critical environments
  • Validated to 10-6 SAL
  • Sterile inner wrapping

Outer Tyvek/polyethylene wrapping minimizes the risk of false contamination, provides fiber-free bacterial barrier and allows for gas exchange. To use, remove the outer wrapping in the non-critical environment, then remove the sterile inner wrapping in the clean area; the included bag may be used to transport plates from your critical environment. Storage temperature: 2 to 8 °C.

Settling plates for active and passive air sampling.

Ordering information: Each pack or case contains a signed, lot-specific Certificate of Analysis.