2X YT Agar Plates with Chloramphenicol 34 and 1% Glucose, Teknova
CA100219-024
: Teknova
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:2YT Agar with 34 µg/mL Chloramphenicol and 1% Glucose
- Size:150 mm
- Environmentally Preferable:
- Cat. no.:CA100219-024
- Supplier No.:Y4285
2X YT is a nutitionally rich medium used for cultivating recombinant strains of Escherichia coli.
- 20 Plates
- Sterile
Contains 1.6% tryptone, 1.0% yeast extract, 0.5% NaCl, 1.5% agar, 34µg/ml Chloramphenicol, 1.0% glucose.
Ambient shipping conditions. Refrigerate to store.