About this item
YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.
- 20 Plates
- Sterile
Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).
Ambient shipping conditions. Store at room temperature or 2–8°C.
8 Options Available Below
Product Details & Documents
YPD is a nutritionally-rich, undefined, non-selective media for the growth and maintenance of Saccharomyces cerevisiae.
- 20 Plates
- Sterile
Ingredients: 1.0% Yeast Extract (10.0 g/L), 2.0% Peptone (20.0 g/L), 2.0% Glucose (20.0g/L), 2.0% Agar (20.0g/L).
Ambient shipping conditions. Store at room temperature or 2–8°C.
Documents
Product Family Options
Product Information
- SizeAvailabilityPrice
- CA100216-446YPD AgarSupplier Number: Y1000
- CA100217-054YPD AgarSupplier Number: Y1001
- CA100216-448YPD Agar Plates, 40 mL/PlatesSupplier Number: Y1030
- CA100218-980YPD Agar plates with Sorbitol and 500 µg/ml ZeocinYPD Agar plates with Sorbitol and 500 µg/ml Zeocin100 mmLog in for availabilityPack of 20$4,976.50Supplier Number: Y1040
- CA100217-508YPD Agar with Sorbitol and 2 mg/mL ZeocinYPD Agar with Sorbitol and 2 mg/mL Zeocin100 mmDiscontinued-Supplier Number: -
- CA100219-702YPD Plates with 1.2 M SorbitolSupplier Number: Y1010
- CA77442-858YPD AgarSupplier Number: Y1001Specifications:New ProductNew Product
- CA77442-856YPD Plates with 1.2 M SorbitolSupplier Number: Y1010Specifications:New ProductNew Product
- CA77442-852YPD Agar plates with Sorbitol and 500 µg/ml ZeocinSupplier Number: Y1040Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



