LB Agar Plates with 150 µg/mL Carbenicillin, Teknova
Catalog # CA200066-622
Supplier: Teknova
CAS Number:
Specifications
- Description:LB Agar with 150 µg/mL Carbenicillin
- Size:245 x 245 mm
- Cat. no.:CA200066-622
- Supplier No.:L6011
Specifications
About this item
Vented Qtray bioassay tray.
- 4 pack
- Sterile
Contains 1% Tryptone, 0.5% yeast extract, 1.0% sodium chloride, 1.5% agar, 150 µg/mL carbenicillin.
Ambient shipping conditions. Refrigerate to store.