About this item
For use with ampicillin resistant strains and strains harboring plasmids such as pBluescript, pGEM, pUC series plasmids. X-gal allows blue/white selection of colonies.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 100 µg/ml ampicillin, 50 µM IPTG, 80 µg/ml X-gal.
Ambient shipping conditions. Refrigerate to store.