LB Agar Plates with Gentamicin 7, Kanamycin 50, Tetracycline 10, Bluo-gal 100 and IPTG, Teknova
Catalog # CA100216-660
Supplier: Teknova
CAS Number:
Specifications
- Description:LB Agar with 7 µg/mL Gentamycin, 50 µg/mL Kanamycin, 10 µg/mL Tetracycline,á100 µg/mL Bluo-gal, and 40 µg/mL IPTG
- Size:100 mm
- Cat. no.:CA100216-660
- Supplier No.:L1919
Specifications
About this item
For use with gentamicin, kanamycin, tetracycline resistant strains.
- Sleeve of 20 plates
- Sterile
Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 7 µg/ml Gentamicin, 50 µg/ml kanamycin, 10 µg/ml tetracycline, 100 µg/ml bluo-gal, and 40 µg/ml IPTG.
Ambient shipping conditions. Refrigerate to store.