Order Entry
Canada
ContactUsLinkComponent
LB Agar Plates with 50 µg/ml Kanamycin and 20 µg/ml Tetracycline, Teknova
LB Agar Plates with 50 µg/ml Kanamycin and 20 µg/ml Tetracycline, Teknova
Catalog # CA100217-390
Supplier:  Teknova
CAS Number:  
LB Agar Plates with 50 µg/ml Kanamycin and 20 µg/ml Tetracycline, Teknova
Catalog # CA100217-390
Supplier:  Teknova
Supplier Number:  L1235
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    LB Agar with 50 µg/mL Kanamycin and 20 µg/mL Tetracycline
  • Size:
    100 mm
  • Environmentally Preferable:
  • Cat. no.:
    CA100217-390
  • Supplier No.:
    L1235

Specifications

About this item

For use with kanamycin and tetracycline resistant strains, and strains harboring plasmids such as pAD22, pUC series plasmids.

  • Sleeve of 20 plates
  • Sterile

Contains 1% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% agar 50 µg/ml kanamycin, 20 µg/ml tetracycline.

Ambient shipping conditions. Refrigerate to store. Protect from light.