Hearts Models And Dissection Bundle
Catalog # 470230-952
Supplier: Avantor
Specifications
- Description:Heart Model and Pig Heart
- Cat. no.:470230-952
Specifications
About this item
Combining A Reference And Your Dissection Specimens
- Buy together and save
Combine a, two part heart, life size heart model with a pail of 10 preserved hearts for dissection. Learn about heart anatomy with the 3B basic heart model. This highly detailed model divides to show details of interior anatomy as well as exterior anatomy. The model has 40 structures identified with numbers that can be matched to the included key. Once your students understand what they will be seeing, use the heart as a reference while they dissect our expertly preserved pig hearts. The preserved hearts of a similar size to the model and can be easily compared for structural identification by the students.