3B Scientific® Torso Skeleton and Stand
470230-946
: Avantor
- Description:Spine and Stand
- Environmentally Preferable:
- Cat. no.:470230-946
Get The Support You Need.
- Buy together and save
- 3-year manufacturers warranty
A Flexible spine with ribs show the interaction of the ribs, spine and associated structures. This spine and rib model contains the following features: Full pelvis and occipital plate, Fully flexible mounting, L3-L4 disc prolapsed, Spinal nerve exits, Cervical vertebral artery, Male pelvis, Movable mounted femur heads. This spine and rib is an anatomically correct model of this section of human anatomy. As part of the bundle a Stand is also included with this model. The stand can be used as a 2 part counter stand with a base for moving around the classroom, or as a wall mounted hanger for display and storage of the model.