Order Entry
Canada
Orders LinkContactUsLinkComponent
PineCAR Racer Basic Car Kit

PineCAR Racer Basic Car Kit

17.7 cm (7"), 4.44 cm (1¾")
Catalog # 470229-848
Supplier:   KIDDER MFG. CO. LTD
Supplier #:  19-8010-00
PineCAR Racer Basic Car Kit
Catalog # 470229-848
PineCAR Racer Basic Car Kit
PineCAR Racer Basic Car Kit

17.7 cm (7"), 4.44 cm (1¾")

Price
15,80 $
Log in for availability
Kit of 1
Product Details & Documents

Individual design kit for pinewood racing.

  • Kit includes wood block, wheels, hubcaps, two lead-free one-piece axles and four lead-free nail-type axles

Wheels and axles should be the same on all cars in a race.

Specifications
  • Length:
    17.7 cm (7")
  • Width:
    4.44 cm (1¾")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.