ZR-96 MagStand, Zymo Research
Catalog # 76005-120
Supplier: Zymo Research
Specifications
- Description:ZR-96 MagStand
- Size:96 well
- Cat. no.:76005-120
Specifications
About this item
The ZR-96 MagStand is a magnetic stand for the separation of magnetic beads in standard 96-well format plates, especially for EZ-96 DNA Methylation MagPrep product line, ZR-96 Genomic DNA MagPrep, Zyppy-96 Plasmid MagBead Miniprep, and Direct-zol-96 MagBead RNA.