Zymoprep™ Yeast Plasmid Miniprep II, Zymo Research
Catalog # 76212-310
Supplier: Zymo Research
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Yeast/Fungi
- Target molecule:Plasmid
- Description:Zymoprep™ Yeast Plasmid Miniprep II
- Size:50 Preps
- Description:Zymoprep™ Yeast Plasmid Miniprep II
- Yield:Typically between 0.01-0.3 ng for most 2 µ based plasmids from 1.5 ml overnight cultures
- Cat. no.:76212-310
- Format:Spin column
Specifications
About this item
The Zymoprep™ Yeast Plasmid Miniprep II used for plasmid isolation from S. pombe.
- Simple and Efficient
- No need for glass beads or phenol
System is ideal for low-copy number and hard to isolate plasmids. Eluted plasmid DNA can be used directly for E. Coli transformation, PCR, and Southern blot analysis.
cerevisiae, C. albicans and S. pombe, and any fungi whose cell walls are susceptible to yeast lytic enzyme lysis.