Purple Broth, Hardy Diagnostics
Supplier: Avantor
Certificates
About this item
For the determination of carbohydrate fermentation by bacteria, especially enteric bacilli and Enterococcus spp
Contains a durham tube for gas detection.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.