ContactUsLinkComponent
LIM Broth, for Group B Strep
LIM Broth, for Group B Strep
  CA76047-150
 :  
undefined
LIM Broth, for Group B Strep
  CA76047-150
 :  L57
 :  
  • Pack type:
    PC tube, optically clear, shatter resistant
  • Description:
    LIM Broth
  • Size:
    20/pack
  • Cat. no.:
    CA76047-150
  • Tube Size:
    13×100 mm
LIM Broth is recommended for the selective enrichment of group B streptococci (Streptococcus agalactiae) from anovaginal specimens from pregnant women.
  • Flat bottom contributes to stability when set on end
  • Leak-proof caps provide security during handling and transportation
  • Robust construction tubes have excellent impact and are shatter resistant