Ion exchange Chromatography Columns, BioPro Series, YMC America
Catalog # 97032-596
Supplier: YMC America,Inc
Specifications
- Ø int.:4.60 mm
- Particle Size:5 µm
- Length:100 mm
- Environmentally Preferable:
- Column Hardware:PEEK
- Cat. no.:97032-596
- Packing Material:SP-F
- Supplier No.:SF00S05-1046WP
Specifications
About this item
YMC-BioPro ion exchange columns are used for the separation of proteins, peptides, and nucleic acids.
- The non-porous particles offer high efficiency and exceptional resolution
YMC-BioPro IEC columns are available in QA and SP chemistries and are based on porous (QA and SP columns) and non-porous (QA-F and SP-F columns) hydrophilic polymer beads with low nonspecific adsorption.