You searched for: Beakers
Graduated and non-graduated beakers allow contents to be efficiently mixed, analyzed, and transported safely. The standard or low-form cylindrical vessels have a reinforced rim typically with a spout for mess-free pouring. With heat and chemical resistance, the containers offer maximum control even under strenuous conditions and procedures. Available in a variety of capacities and manufacturing materials, the laboratory beakers may possess additional attributes including color-coding, non-slip handles, and no-drip spouts.
VWR® Heavy-Duty Low Form Beakers
Supplier: VWR International
These beakers are mechanically stronger than standard beakers and are designed for longer life with increased user safety.
Expand 6 Items
VWR® Beakers without Handle, PP
Supplier: VWR International
VWR® polypropylene beakers, which come in a natural color, have a pouring spout and graduations printed directly on the beakers.
Expand 9 Items
VWR® TPX™ Beakers with Handle
Supplier: VWR International
VWR® TPX™ Beakers, which are crystal-clear in color, have a handle, pouring spout and graduations printed directly on the beakers.
Expand 6 Items
Kartell® Graduated Beakers, Low Form, PMP
Supplier: VWR International
Crystal-clear beakers feature excellent chemical resistance and a large spout for efficient pouring.
Expand 8 Items
Standard-Grade Glass Beaker
Supplier: VWR International
Our high-quality borosilicate beakers meet ASTM standards and are a great value. With uniform wall thickness, white enamel, double-graduated metric scale, and a marking spot. Beakers can be heated and used with acids, salt solutions, and organic solvents.
Expand 12 Items
VWR®, Graduated Beakers, Polypropylene
Supplier: VWR International
Beakers feature a pouring spout for quick and easy dispensing.
Expand 8 Items
VWR®, Griffin Beakers, Polypropylene
Supplier: VWR International
Translucent walls and molded graduations make it easy to prepare, mix, and measure solutions within the beaker.
Expand 9 Items
VWR® Standard Standard-Grade Beakers
Supplier: VWR International
A Great Value!
Expand 13 Items
VWR® Beakers with Handle, PP
Supplier: VWR International
VWR® polypropylene beakers, which come in a natural color, have a handle, pouring spout and graduations printed directly on the beakers.
Expand 7 Items
Graduated Polypropylene Pitchers
Supplier: VWR International
These pitchers are made with shatter-proof polypropylene.
Expand 6 Items
VWR® Low Form Heavy Duty Beakers
Supplier: VWR International
Our high-quality borosilicate beakers meet ASTM standards at a great value.
Expand 6 Items
VWR® Heavy-Duty-Grade Beakers
Supplier: VWR International
0.5 mm Thicker Wall Than Standard Grade Beakers
Expand 5 Items
VWR® Low Form Griffin Beakers
Supplier: VWR International
These low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass.
Expand 15 Items
VWR®, Beaker Cups, Polystyrene
Supplier: VWR International
Beaker cups are designed for use with various automated procedures and as an economical, general-purpose sample cup.
Expand 6 Items
VWR® Berzelius Tall Form Beakers
Supplier: VWR International
Our new high-quality borosilicate Berzelius beakers meet ASTM standard and are a great value.
Expand 7 Items
VWR®, Beakers, PTFE
Supplier: VWR International
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
Expand 13 Items
VWR®, Graduated Beakers with Handle, Low Form, Polypropylene
Supplier: VWR International
Graduated beakers are robust, with a non-drip spout and ridged thumb-grip on handle.
Expand 5 Items
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Supplier: VWR International
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
Expand 6 Items
VWR® Thermotech® Beakers, PTFE
Supplier: VWR International
These unique beakers are a combination of a pure PTFE body with a stabilized PTFE carbon base to form a single molding.
Expand 3 Items
VWR® Tall Form Beaker
Supplier: VWR International
High Quality at a Great Value
Expand 6 Items
VWR® TPX™ Beakers without Handle
Supplier: VWR International
VWR® TPX™ Beakers, which are crystal-clear in color, have a pouring spout and graduations printed directly on the beakers.
Expand 8 Items
Beaker with Dual Temp Strip, 6.5 oz., Therapak
Supplier: Therapak, LLC
This wide-mouth cup is ideal for initial collection of urine samples and subsequent pour off into transport vials.
Expand 1 Items
Beaker Mug, Graduated, Ace Glass Incorporated
Supplier: Ace Glass
This is a 400mL heavy-wall, graduated beaker mug
Expand 1 Items
Beakers, Quartz, Ace Glass Incorporated
Supplier: Ace Glass
This low form, quartz beaker has a pouring spout
Expand 7 Items
Beakers, Jacketed, Ace Glass Incorporated
Supplier: Ace Glass
This jacketed beaker has one upper and one lower Ace-Thred connection on opposite sides.
Expand 9 Items
Nalgene® PFA Beakers with Handles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Nalgene® PFA Beakers with Handles feature extraordinary chemical resistance and can be used repeatedly for high-purity applications
Expand 2 Items
Nalgene® Griffin Beakers, Low Form, Teflon® PFA, Thermo Scientific
Supplier: Thermo Fisher Scientific
Beakers offer excellent chemical resistance for aggressive or high-purity chemicals and solutions.
Expand 4 Items
SP Bel-Art Large Volume Beakers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
These graduated beakers and pitchers feature flared rims and spouts for quick, easy pouring when large volumes are handled.
Expand 2 Items
SP Bel-Art Graduated Griffin Low-Form Beakers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Griffin low-form polypropylene plastic beakers are translucent, chemical resistant, and will stack without sticking together.
Expand 9 Items
Beakers, Copper, Thermo Scientific
Supplier: Thermo Scientific Chemicals
These beakers are made up of copper.