About this item
Rapid, sensitive and precise probe-based qPCR detection and quantitation of RNA targets.
In the Luna Universal One-Step Probe RT-qPCR Kit, Hot Start Taq DNA Polymerase is combined with a novel WarmStart-activated reverse transcriptase, allowing dual control of enzyme activity via reversible, aptamer-based inhibition.
The Luna Universal Probe One-Step Reaction Mix is supplied at 2X concentration and contains Hot-Start Taq DNA Polymerase, dNTPs, and all required buffer components. It is formulated with a unique passive reference dye that is compatible across a variety of instrument platforms, including those that require a high or low ROX reference signal. The Reaction Mix also features dUTP for carryover prevention and a non-fluorescent visible dye for monitoring reaction setup. This visible dye does not overlap spectrally with fluorophores commonly used in qPCR and does not interfere with real-time detection.
The Luna WarmStart RT Enzyme Mix is supplied at 20X concentration and contains Luna WarmStart Reverse Transcriptase as well as Murine RNase Inhibitor to aid in preventing RNA degradation. It is compatible with various RNA sample types (total RNA, poly(A)-RNA, etc.) and sources.
This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling, providing added security for setting up reactions at room temperature. The engineered WarmStart Luna Reverse Transcriptase also possesses higher thermostability than many other RTs, allowing an optimal reaction temperature of 55 °C. For difficult targets/templates, higher RT step temperatures of up to 60 °C can be used without compromising Luna performance.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:RT-qPCR probe
- One step:Yes