ContactUsLinkComponent
VWR® TraceableLIVE® Wi-Fi Data Logging LN₂ Thermometers with Remote Notification
undefined
VWR® TraceableLIVE® Wi-Fi Data Logging LN₂ Thermometers with Remote Notification
  • Catalog No:
  • 76047-268
  • 76047-270
  • Interface:
  • WiFi
  • WiFi
  • Weight:
  • 113 g
  • 113 g
  • W×D×H:
  • 108×70×19 mm (4.25×2.75×0.75")
  • 108×70×19 mm (4.25×2.75×0.75")
  • Batteries:
  • 4 AAA Alkaline battery
  • 4 AAA Alkaline battery
Frequently Bought Together