FreeZone® Freeze Dry Glassware Kit, Labconco
89428-604
: Labconco
- Description:FreeZone® Freeze Dry Glassware Kit
- Cat. no.:89428-604
- W×D×H:12×10×6"
- Weight:5.0 lbs.
- Supplier No.:7540100
Replacement glassware kit for FreeZone® freeze dry systems.
: Includes one each 600 ml Complete Fast-Freeze Flask, one each 300 ml Complete Fast-Freeze Flask, one each 3/4" SST Straight Adapter, and one each 3/4" SST 45° Bend Adapter.