BD Taxo™ X Factor Discs, BD Diagnostic Systems
Catalog # CA-90002-554
Supplier: BD
CAS Number:
Specifications
- Description:BD Taxo™ X Factor Discs for Haemophilus species
- Size:50 discs
- Cat. no.:CA-90002-554
- Supplier No.:231729
Specifications
About this item
BD Taxo™ X Factor Discs are used for differentiating Haemophilus species.
These paper discs are round, brown 1/4” with “x” printed on both sides.