Ni-NTA His-Bind® Superflow™ Resin, EMD Millipore
Supplier: MilliporeSigma
Certificates
About this item
Ni-NTA HisBind® Superflow™ Resin is a medium-pressure compatible version of the Ni-NTA HisBind® Resin
- Binding capacity of 5 - 50 mg
- Storage at 2 to 8 °C
Compatible with FPLC or gravity-flow applications.
It has all the same characteristics, except the agarose matrix has a higher level of crosslinking for higher bead rigidity.
Specifications
- Storage temperature:10 mL
- Binding capacity:5 - 50 mg/mL
Specifications
Related Information
Frequently Bought Together