Universal Wastewater Standard, NSI Lab Solutions
Catalog # NSQCI-084
Supplier: SPEX CERTIPREP LLC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Universal Wastewater Standard
- Size:4×1 L
- Ammonia nitrogen:7.50 mg/L
- BOD:198 mg/L (BOD₅)
163 mg/L (CBOD) - Nitrate nitrogen:3.00 mg/L
- ortho-Phosphate:2.50 mg/L
- Total suspended solids (tss):100 mg/L "
- COD:292 mg/L
- Conductivity:600 umhos
- TDS:450 mg/L
- TOC content:121 mg/L
- pH:7.2 units
- Total solids:725 mg/L
- Cat. no.:NSQCI-084
- No. of Analytes:12
- Supplier No.:QCI-084
Specifications
About this item
Universal Wastewater Standard is a Ready-to-use glass bottle
- Stable and remarkably easy to use
- Shake and pour an aliquot
- Parameters are derived from 1:1 Glucose:Glutamic acid at 150 mg/L each
Designed to reduce the number of standards used, this standard is certif ed for 12 conventional pollutants. Works with all listed EPA approved methods.
It is provided as a sterile solution which can be stored at room temperature prior to opening. Once opened, it should be stored tightly capped at 4°C.