You searched for: Retort Stands
Retort stands to hold burettes, flasks, and other labware during procedures are available at VWR. Sets include both stainless steel rod and base, constructed of your choice of plastic or cast iron. A polypropylene retort stand is offered for severe corrosive conditions. Bases of cast iron coated with enamel are featured in rectangular, tripod, U-shaped and A-shaped options. Different rod lengths for the support of different diameters of labware are also supplied through VWR, as well as retort rings and clamps.
VWR® Talon® Support Stand, Cast Iron, U-Shaped Base
Supplier: VWR International
Support stand base is constructed of cast iron with black enamel finish.
Expand 1 Items
VWR® Talon® Support Plates
Supplier: VWR International
Designed to hold hotplate/stirrers and other apparatus to lab frames or ring stands.
Expand 3 Items
VWR® Talon® Support Stands
Supplier: VWR International
Support stands are made entirely of Type 303 stainless steel and can accommodate vessels up to 45.7 cm (18") in diameter within the sturdy U-shaped base.
Expand 6 Items
A-Frame Base
Supplier: VWR International
This cast iron base features threads for rods sized 1/2 - 13.
Expand 2 Items
VWR® Talon® Gas Cylinder Stand
Supplier: VWR International
Permits convenient, safe cylinder transfer and upright support.
Expand 1 Items
Rectangular Bases
Supplier: VWR International
Essential for Any Chemistry or Physics Laboratory
Expand 3 Items
VWR® Talon® Support
Supplier: VWR International
Multi-purpose support was originally designed to permit cylindrical vessels up to 305mm (12") in diameter to sit close to the upright rod.
Expand 1 Items
VWR® Talon® Heavy-Duty Support Stands
Supplier: VWR International
Heavy-duty support stands permit cylindrical vessels up to 30.5 cm (12") in diameter to sit close to the support rod.
Expand 6 Items
Rod and Base Assembly
Supplier: VWR International
This assembly is handy for multiple lab applications.
Expand 3 Items
VWR® Talon® Adjustable Cylinder Stand
Supplier: VWR International
Heavy-duty, cast aluminum stand with adjustable L-brackets holds cylinders from 15.2 – 23.5 cm (6 – 91/4") in diameter.
Expand 1 Items
Funnel Support
Supplier: VWR International
With iron clamp for support rods of any size to 5/8". The four holes are beveled and slotted and are spaced at 41/2" center distance.
Expand 1 Items
High-Low Alcohol Burner Stand
Supplier: VWR International
High-Low Alcohol Burner Stand
Expand 2 Items
Tripod Bases
Supplier: VWR International
These heavy cast iron bases provide a three-point support for stability.
Expand 3 Items
Fun Stand
Supplier: VWR International
Ensure accuracy in all your experiments.
Expand 1 Items
Funnel Support with Base
Supplier: VWR International
Made from fine, oil finished hardwood designed to support up to four funnels. The height is adjustable and the holes are slotted for easy placement of funnels. The base measures 11 x 38 cm.
Expand 1 Items
VWR® Talon® Aluminum Support Plate
Supplier: VWR International
Talon® Aluminum Support Plate fits standard lattice lab frames using VWR® Talon® connectors 21572-501, 21572-556, or 60097-055.
Expand 1 Items
T-Nesting Support Bases
Supplier: VWR International
Specially designed to provide excellent stability, these bases are shaped with legs that slide freely under one another for compact storage or set up.
Expand 1 Items
VWR® Talon® Cylinder Stand
Supplier: VWR International
Cast aluminum stand hinges open and has four thumbscrews to tightly grip cylinders from 20.3 – 22.9 cm (8 – 9") in diameter.
Expand 1 Items
VWR® Talon® Support Stands, Cast Iron, Rectangular Base
Supplier: VWR International
Support stand bases are constructed of cast iron with black enamel finish.
Expand 4 Items
VWR® Talon® Support Stand, Cast Iron, Tripod Base
Supplier: VWR International
Support stand base is constructed of cast iron with black enamel finish.
Expand 1 Items
VWR® Talon® Double Buret Holder and Support Stand
Supplier: VWR International
Double buret clamp holds any size buret from micro to 100 mL capacity.
Expand 6 Items
LaboPlast Funnel Stands for Two or Four Funnels, Bürkle
Supplier: BURKLE INC
Sturdy and stable funnel stand, resistant to chemicals, shatter-proof and with continuously variable height adjustment.
Expand 2 Items
Labjaws FootPlate, Ace Glass Incorporated
Supplier: Ace Glass
The Labjaws Footplate holds Flexaframe poles
Expand 1 Items
Photochemical Reactor Stand and Stand Inserts, Ace Glass Incorporated
Supplier: Ace Glass
Sturdy aluminum, powder coated stand for use with cylindrical reactors.
Expand 9 Items
Bench Top Reactor Stand, Ace Glass Incorporated
Supplier: Ace Glass
This newer bench reactor stand assembly accommodates all types of glass bench filter and pressure reactors.
Expand 1 Items
SP Bel-Art Lattice Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Lattice support stand is a space-saving option for the benchtop.
Expand 1 Items
SP Bel-Art Compact Support Stand, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This non-corrosive support stand with stainless steel mounting rod provides modern advantages over metal ring stands.
Expand 1 Items
Plate Support Shelf, Ace Glass
Supplier: Ace Glass
Adjustable cast alloy, black epoxy coated support shelf
Expand 2 Items
Support Stand, 'C' Shape, All Stainless Steel, Ace Glass Incorporated
Supplier: Ace Glass
An all-stainless-steel support stand with a heavy, six pound base