KIMBLE® Micro Soxhlet Extraction Apparatus, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Soxhlet-type extraction apparatus is for use with extraction thimble, which has a coarse porosity.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought