Order Entry
Canada
ContactUsLinkComponent
Methyl Green
Methyl Green
Catalog # 470229-146
Supplier:  Ward's Science
CAS Number:  
undefined
Methyl Green
Catalog # 470229-146
Supplier:  Ward's Science
Supplier Number:  AD-MM0410-5G
CAS Number:  

Specifications

  • Description:
    Methyl Green
  • Size:
    5 g
  • Chemical/Physical Specifications:
    Powder
  • Cat. no.:
    470229-146
  • Grade:
    Laboratory

Specifications

About this item

CAS Number: 7114-03-6
Formula Weight: 608.78
Formula: C27H35Cl2N3·ZnCl2
Density (g/mL): >1
Boiling Point (°C): Decomposes
Solubility: Moderately soluble in water
Shelf Life (months): 36
Storage: Green

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.