Specifications
- Maximum Temperature:1230 °C (2250 °F)
- Model:AC1228/PM6
- Power:6.2 KW
- Exterior W×D×H:25×28×34"
- Interior W×D×H:12×20×8"
- Cat. no.:80701-372
- Electrical:240 VAC, 26 A
- Weight:300 lbs.
- Supplier No.:FAC1228/PM6
Specifications
About this item
Model AC1228/PM6 is a heat treating furnace used for treating metals with a maximum temperature of up to 2250°F.
- Features a Watlow PM6 controller with double display single set point, no timer
- Chamber is constructed of firebrick of the best quality
The Assaying option consists of the Cress clean air vent system, along with embedded (sealed) heating elements. Element embedding seals the element wires in the porcelain plates and helps protect them from contaminants that can cause premature failure. The Cress clean air vent system draws a small amount of fresh air through the furnace chamber to remove contaminants, this is mixed with a large amount of room air and expelled out through a standard 4" metal duct.
It measures 2.5” and it is backed with 1.5” of 1800°F block insulation. The furnace shell is constructed of 18 GA sheet metal. The door opens up away from the user. There is a rope seal on the door that provides a better seal against the furnace. The door mechanism uses two heavy duty springs that keep the door safely closed or open.