Order Entry
ContactUsLinkComponent
Pierce Easy-Titer® IgM Assay Kits, Thermo Scientific
Pierce Easy-Titer® IgM Assay Kits, Thermo Scientific
Catalog #: CAPI23315
Supplier:  Thermo Scientific
CAS Number:  
Pierce Easy-Titer® IgM Assay Kits, Thermo Scientific
Catalog #: CAPI23315
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Human IgM Kit
  • Detection range:
    15–300 ng/mL
  • Intra- and interassay variation:
    \<5%
  • Cat. no.:
    CAPI23315
  • No. of Assays/Kit:
    96 assays
  • Supplier No.:
    23315

Specifications

About this item

Easy-Titer® IgM Assay Kit is a simple, mix-and-read assay that allows quick and accurate determination of concentrations from 15 to 500 ng/mL of IgM

  • Faster and easier than ELISA
  • Sensitive, requires small sample volume
  • No special equipment needed
  • Highly reproducible results

The assay procedure uses monodispersed polystyrene beads that are coated with anti-human IgM antibodies and absorb light at 340 and 405 nm. When the beads are mixed with a sample containing human IgM, they aggregate, causing decreased absorption of light and, therefore, low IgM concentrations yield high absorbance values and high IgM concentrations yield low absorbance values. The decrease in absorption is proportional to IgM concentration and a standard curve can be generated to accurately quantify IgM levels in serum or citrated plasma samples.

Delivery: Kit include sufficient components for 96 tests. An IgM standard must be purchased separately. Each kit contains goat Anti-Human IgM Coated polystyrene beads (2 mL), Dilution buffer (30 mL), and blocking reagent (15 mL).
Customers Who Bought This Also Bought