Ezwaste™ HDPE Containers, UN/DOT-Rated, Foxx Life Sciences
Catalog # 76018-504
Supplier: VWR International
Specifications
- Capacity:13.5 L
- Description:HDPE Containers, UN/DOT-Rated, Bulk Packaging
- Cat. no.:76018-504
Specifications
About this item
Containers allow for the safe and effective collection of waste in the laboratory.
- UN and DOT certified for transport and shipping while full
- Cost-effective for biotech, pharma, and teaching labs alike
- Durable product with handle for easy pouring
The EZwaste™ UN/DOT system is designed to be an affordable solution for liquid waste disposal. Containers were made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is supplied with a 70S cap.