LaboPlast® Bürkle Thread Adapter, 3/4" Inflowing, Coarse Thread , Tri-Sure 2" Outflowing, Bürkle
Catalog # 76045-418
Supplier: BURKLE INC
Specifications
- Color:Green/White
- Description:LaboPlast® Thread Adapter
- Material:PP/PE
- Cat. no.:76045-418
- Thread Size:Outflowing: Coarse Thread, Tri-Sure 2"
Inflowing: 3/4"
Specifications
About this item
Designed for use with pumps, these adapters feature a coarse, Tri-Sure 2" thread on the outflowing side, and a ³/₄" thread on the inflowing side.
Stopcock spigots, ball valves, and other accessories can be connected to most commercially available containers with LaboPlast® thread adapters.
Packaging: One adapter per package.